Learn More
BIOHIPPO Human DEFB103B Microsample
SDP
Catalog No. 5000315941
Encompass
Human DEFB103B Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of DEFB103B in Serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 11 7 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human DEFB103B Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human DEFB103B Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human DEFB103B biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15941 Supplier BIOHIPPO Supplier No. BHE15211041
Human DEFB103B Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of DEFB103B in Serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 11 7 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human DEFB103B Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human DEFB103B Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human DEFB103B biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color