Learn More
Biomatik Corporation Human Defensin Alpha 4, Corticostatin (DEFa4) ELISA Kit
Supplier:  Biomatik Corporation EKU03651
Encompass_Preferred
Human Defensin Alpha 4, Corticostatin (DEFa4) ELISA Kit for detecting DEFa4 in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P12838. Gene ID: 1669. Target Synonyms: DEF4; HP-4; HNP-4; Neutrophil defensin 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9016