Learn More
Biomatik Corporation Human Defensin Alpha 5, Paneth Cell Specific (DEFa5) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU10921
Encompass_Preferred
Human Defensin Alpha 5, Paneth Cell Specific (DEFa5) High-sensitive ELISA Kit for detecting DEFa5 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q01523. Gene ID: 1670. Target Synonyms: DEF-A5; DEF5; HD-5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5638