Learn More
Biomatik Corporation Human Defensin Alpha 6, Paneth Cell Specific (DEFa6) ELISA Kit
Supplier:  Biomatik Corporation EKU03654
Encompass_Preferred
Human Defensin Alpha 6, Paneth Cell Specific (DEFa6) ELISA Kit for detecting DEFa6 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 46.88-3,000pg/ml. Sensitivity: 19.17pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q01524. Gene ID: 1671. Target Synonyms: DEF6; Defensin-6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9015