Learn More
Biomatik Corporation Human Defensin Beta 119 (DEFb119) ELISA Kit
Supplier:  Biomatik Corporation EKU09416
Encompass_Preferred
Human Defensin Beta 119 (DEFb119) ELISA Kit for detecting DEFb119 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.125ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8N690. Gene ID: 245932. Target Synonyms: DEFB120; DEFB-19; DEFB-20. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6475