Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human Dendritic Cell Cytokine Package, PeproTech®

Catalog No. HDC
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
HDC 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. HDC Supplier Gibco™ Supplier No. HDC
  • $1,036.00
    $777.65 / Each of 1

    Save $258.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Human Dendritic Cell Cytokine Package contains the key components required for the culturing of human dendritic cells.

IL-4 is a pleiotropic cytokine that regulates diverse T and B cell responses including cell proliferation, survival, and gene expression. Produced by mast cells, T cells, and bone marrow stromal cells, IL-4 regulates the differentiation of naive CD4+ T cells into helper Th2 cells.

GM-CSF is a hematopoietic growth factor that stimulates the development of neutrophils and macrophages and promotes the proliferation and development of early erythroid megakaryocytic and eosinophilic progenitor cells.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Functional Assay
Product Type Dendritic Cell Cytokine Package
Quantity 1 Kit
Content And Storage 100 μg Human IL-4 (Cat. No. 200-04-100 UG), 100 μg Human GM-CSF (Cat. No. 300-03-100 UG)
Species Human
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.