Learn More
Sino Biological Human DENR PAb 100ul (Sino Biological)
Supplier:  Sino Biological 203057T32100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human DENR fragment, and purified by antigen affinity chromatography.
Catalog No. 50-237-1812