Learn More
Biomatik Corporation Human Density Enhanced Phosphatase 1 (DEP1) ELISA Kit
Supplier:  Biomatik Corporation EKU06946
Encompass_Preferred
Human Density Enhanced Phosphatase 1 (DEP1) ELISA Kit for detecting DEP1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.117ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q12913. Gene ID: 5795. Target Synonyms: CD148; PTPR-J; PTPRJ; HPTPeta; R-PTP-ETA; SCC1; Protein-tyrosine phosphatase eta; Density-enhanced phosphatase 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7648