Learn More
Biomatik Corporation Human Deoxycytidine Kinase (DCK) ELISA Kit
Supplier:  Biomatik Corporation EKU03689
Encompass_Preferred
Human Deoxycytidine Kinase (DCK) ELISA Kit for detecting DCK in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P27707. Gene ID: 1633. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4662