Learn More
American Research Products Inc Human deoXyribonuclease I DNa
Catalog No. 5000329229
Encompass
Human deoXyribonuclease I DNase-I ELISA Kit from CUSABIO Detection Range 1 56 ng/mL-100 ng/mL Sensitivity 0 39 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29229 Supplier American Research Products Inc Supplier No. CSBE09068H
Human deoXyribonuclease I DNase-I ELISA Kit from CUSABIO Detection Range 1 56 ng/mL-100 ng/mL Sensitivity 0 39 ng/mL Sample serum plasma tissue homogenates Method Sandwich