Learn More
Biomatik Corporation Human Deoxyribonuclease II (DNASEII) ELISA Kit
Supplier:  Biomatik Corporation EKU11989
Encompass_Preferred
Human Deoxyribonuclease II (DNASEII) ELISA Kit for detecting DNASEII in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.117ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O00115. Gene ID: 1777. Target Synonyms: DNase-II; DNase2; DNASE2A; DNASE2B; DRN2; DNL; DNL2; Deoxyribonuclease II Alpha; Acid DNase; Lysosomal DNase II. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6117