Learn More
Biomatik Corporation Human Deoxyribonuclease X (DNASEX) ELISA Kit
Supplier:  Biomatik Corporation EKU03694
Encompass_Preferred
Human Deoxyribonuclease X (DNASEX) ELISA Kit for detecting DNASEX in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P49184. Gene ID: 1774. Target Synonyms: DNASE1L1; DNAS1L1; XIB; DNase-X; DNL1L; Deoxyribonuclease I-Like 1; Muscle-specific DNase I-like. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4886