Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Dermatopontin/TRAMP ELISA Kit

Catalog No. EH151RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH151RBX5 5 x 96 Tests
EH151RBX10 10 x 96 Tests
EH151RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EH151RBX5 Supplier Invitrogen™ Supplier No. EH151RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

Human Dermatopontin/TRAMP quantitates human Dermatopontin/TRAMP in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Dermatopontin/TRAMP.

Dermatopontin is an extracellular matrix protein with possible functions in cell-matrix interactions and matrix assembly. The protein is found in various tissues and many of its tyrosine residues are sulphated. Dermatopontin is postulated to modify the behavior of TGF-beta through interaction with decorin.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q07507
Assay Range 0.43-100 ng/mL
Assay Sensitivity 0.43 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 1810032B19Rik; 22 kDa extracellular matrix protein; 5033416F05Rik; Dermatan sulfate proteoglycan-associated protein 22K; dermatopontin; Dpt; Early quiescence protein 1; early quiescence protein-1; Eq1; EQ-1; hypothetical protein LOC555282; si:dkey-98p3.2; similar to Dermatopontin precursor (Tyrosine-rich acidic matrix protein) (TRAMP); TRAMP; Tyrosine-rich acidic matrix protein; zgc:1366131810032B19Rik; 22 kDa extracellular matrix protein; 5033416F05Rik; Dermatan sulfate proteoglycan-associated protein 22K; dermatopontin; Dpt; Early quiescence protein 1; early quiescence protein-1; Eq1; EQ-1; hypothetical protein LOC555282; si:dkey-98p3.2; similar to Dermatopontin precursor (Tyrosine-rich acidic matrix protein) (TRAMP); TRAMP; Tyrosine-rich acidic matrix protein; zgc:136613;
Gene ID (Entrez) 1805
Gene Symbol DPT
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target DPT
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.