Learn More
BIOHIPPO Human DES Microsample
SDP
Catalog No. 5000313906
Encompass
Human DES Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of DES in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 06 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human DES Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human DES Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human DES biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13906 Supplier BIOHIPPO Supplier No. BHE15210589
Human DES Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of DES in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 06 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human DES Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human DES Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human DES biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color