Learn More
Biomatik Corporation Human Desmin (Des) ELISA Kit
Supplier:  Biomatik Corporation EKU03701
Encompass_Preferred
Human Desmin (Des) ELISA Kit for detecting Des in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.122ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P17661. Gene ID: 1674. Target Synonyms: CMD1I; CSM1; CSM2; Intermediate Filament Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6929