Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Desmoglein-3 ELISA Kit

Catalog No. EH155RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH155RBX5 5 x 96 Tests
EH155RB 96 Tests
EH155RBX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EH155RBX5 Supplier Invitrogen™ Supplier No. EH155RBX5
Add to Cart
Add to Cart

ELISA

Human Desmoglein-3 quantitates human Desmoglein-3 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Desmoglein-3.

Desmoglein 3 (Dsg3), also known as Cadherin family member 6 (CDHF6), is a member of the desmosomal cadherin family and plays a critical role in cell-cell adhesion. It is predominately expressed in stratified squamous epithelium, including the skin, oral mucosa, and esophagus. Desmoglein 3 has been characterized as the auto-antigen of pemphigus vulgaris (PV), a life-threatening autoimmune disease. Binding of anti-Desmoglein 3 autoantibodies causes the loss of cell-cell adhesion, which clinically manifests as blisters on the skin and oral mucosa, and histologically as suprabasilar acantholysis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P32926
Assay Range 34-8000 pg/mL
Assay Sensitivity 34 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 130 kDa pemphigus vulgaris antigen; 130 kDa pemphigus vulgaris antigen homolog; bal; balding; Cadherin family member 6; CDHF6; desmoglein 3; desmoglein-3; DKFZp686P23184; Dsg3; PVA130 kDa pemphigus vulgaris antigen; 130 kDa pemphigus vulgaris antigen homolog; bal; balding; Cadherin family member 6; CDHF6; desmoglein 3; desmoglein-3; DKFZp686P23184; Dsg3; PVA;
Gene ID (Entrez) 1830
Gene Symbol Dsg3
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target Desmoglein 3
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.