Learn More
Biomatik Corporation Human Destrin (DSTN) ELISA Kit
Supplier:  Biomatik Corporation EKU03713
Encompass_Preferred
Human Destrin (DSTN) ELISA Kit for detecting DSTN in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P60981. Gene ID: 11034. Target Synonyms: ADF; ACTDP; Actin Depolymerizing Factor; Actin Binding Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6078