Learn More
Sino Biological Anti-DHRS2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 102488T04100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human DHRS2, and purified by antigen affinity chromatography.
Catalog No. 50-240-8919