Learn More
Sino Biological Human DHRS9 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 15253T52100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human DHRS9 / 3-alpha-HSD (rh DHRS9 / 3-alpha-HSD; Catalog#15253-H07E; NP_005762.2; Arg18-Val319). DHRS9 / 3-alpha-HSD specific IgG was purified by Human DHRS9 / 3-alpha-HSD affinity chromatography.
Catalog No. 50-237-0654