Learn More
Biomatik Corporation Human Diacylglycerol Kinase Alpha (DGKa) ELISA Kit
Supplier:  Biomatik Corporation EKU12200
Encompass_Preferred
Human Diacylglycerol Kinase Alpha (DGKa) ELISA Kit for detecting DGKa in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.053ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P23743. Gene ID: 1606. Target Synonyms: DGK-A; DAGK; DAGK1; DGK-alpha; 80 kDa diacylglycerol kinase; Diglyceride kinase alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8008