Learn More
Biomatik Corporation Human Diacylglycerol-O-Acyltransferase Homolog 2 (DGAT2) ELISA Kit
Supplier:  Biomatik Corporation EKU03719
Encompass_Preferred
Human Diacylglycerol-O-Acyltransferase Homolog 2 (DGAT2) ELISA Kit for detecting DGAT2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.33ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q96PD7. Gene ID: 84649. Target Synonyms: ARAT; Acyl-CoA retinol O-fatty-acyltransferase; Diglyceride acyltransferase 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4437