Learn More
Biomatik Corporation Human Dicarbonyl/L-Xylulose Reductase (DCXR) ELISA Kit
Supplier:  Biomatik Corporation EKU10791
Encompass_Preferred
Human Dicarbonyl/L-Xylulose Reductase (DCXR) ELISA Kit for detecting DCXR in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.2-2,000pg/ml. Sensitivity: 11.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q7Z4W1. Gene ID: 51181. Target Synonyms: KIDCR; DCR; SDR20C1; Short Chain Dehydrogenase/Reductase Family 20C,Member 1; Carbonyl reductase II; Kidney dicarbonyl reductase; Sperm surface protein P34H. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7169