Learn More
Biomatik Corporation Human Dickkopf Related Protein 2 (DKK2) ELISA Kit
Supplier:  Biomatik Corporation EKU03732
Encompass_Preferred
Human Dickkopf Related Protein 2 (DKK2) ELISA Kit for detecting DKK2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5,000pg/ml. Sensitivity: 28pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9UBU2. Gene ID: 27123. Target Synonyms: Dickkopf Homolog 2 (Xenopus Laevis). Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7676