Learn More
Biomatik Corporation Human Dipeptidase 2 (DPEP2) ELISA Kit
Supplier:  Biomatik Corporation EKU03748
Encompass_Preferred
Human Dipeptidase 2 (DPEP2) ELISA Kit for detecting DPEP2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9H4A9. Gene ID: 64174. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9010