Learn More
Biomatik Corporation Human Dipeptidase 3 (DPEP3) ELISA Kit
Supplier:  Biomatik Corporation EKU03749
Encompass_Preferred
Human Dipeptidase 3 (DPEP3) ELISA Kit for detecting DPEP3 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.105ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9H4B8. Gene ID: 64180. Target Synonyms: MBD3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4517