Learn More
Biomatik Corporation Human Dipeptidyl Peptidase 10 (DPP10) ELISA Kit
Supplier:  Biomatik Corporation EKU03750
Encompass_Preferred
Human Dipeptidyl Peptidase 10 (DPP10) ELISA Kit for detecting DPP10 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.116ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8N608. Gene ID: 57628. Target Synonyms: DPL2; DPPY; DPRP3; Dipeptidylpeptidase 10 (Non-Functional) ; Dipeptidyl peptidase IV-related protein 3; Dipeptidyl peptidase X; Dipeptidyl peptidase-like protein 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6186