Learn More
Biomatik Corporation Human Dipeptidyl Peptidase 6 (DPP6) ELISA Kit
Supplier:  Biomatik Corporation EKU03751
Encompass_Preferred
Human Dipeptidyl Peptidase 6 (DPP6) ELISA Kit for detecting DPP6 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.08ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P42658. Gene ID: 1804. Target Synonyms: DPPX; Dipeptidylpeptidase 6; Dipeptidyl aminopeptidase-like protein 6; Dipeptidyl aminopeptidase-related protein; Dipeptidyl peptidase IV-like protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5368