Learn More
Sino Biological Human DKK-1 Matched ELISA Antibody Pair Set 15 Plates
Catalog No. 5000786399
Encompass_Preferred
The Human DKK1 / Dkk-1 ELISA Pair Set is for the quantitative determination of Human DKK1 / Dkk-1.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human DKK1 / Dkk-1 coated on a 96-well plate. Standards and samples are added to the wells, and any Human DKK1 / Dkk-1 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-Human DKK1 / Dkk-1 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human DKK1 / Dkk-1 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-86399 Supplier Sino Biological Supplier No. SEKA1017015
The Human DKK1 / Dkk-1 ELISA Pair Set is for the quantitative determination of Human DKK1 / Dkk-1.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human DKK1 / Dkk-1 coated on a 96-well plate. Standards and samples are added to the wells, and any Human DKK1 / Dkk-1 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-Human DKK1 / Dkk-1 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human DKK1 / Dkk-1 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.