Learn More
Sino Biological Human DKK-3 Matched ELISA Antibody Pair Set 5 Plates
Catalog No. 5000785950
Encompass_Preferred
The Human DKK3 ELISA Pair Set is for the quantitative determination of Human DKK3.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human DKK3 coated on a 96-well plate. Standards and samples are added to the wells, and any Human DKK3 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Human DKK3 polyclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human DKK3 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-85950 Supplier Sino Biological Supplier No. SEKA102035
The Human DKK3 ELISA Pair Set is for the quantitative determination of Human DKK3.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human DKK3 coated on a 96-well plate. Standards and samples are added to the wells, and any Human DKK3 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Human DKK3 polyclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human DKK3 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.