Learn More
Sino Biological Anti-DKK-1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10170T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human DKK-1 (rh DKK-1; Catalog#10170-H08H; NP_036374.1; Met2-His266). DKK-1 specific IgG was purified by Human DKK-1 affinity chromatography.
Catalog No. 50-241-2347