Learn More
Sino Biological Recombinant Human DKK-1 Protein (hFc Tag)
Supplier:  Sino Biological 10170H02H100
Encompass_Preferred
A DNA sequence encoding the human DKK1 (NP_036374.1) (Met2-His266) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-255-5445