Learn More
Sino Biological Human DLG4 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 200060T08100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human DLG4 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-237-1421