Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human DLK1 Synthetic Peptide

Catalog No. PEP1475 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP1475 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP1475 Supplier Invitrogen™ Supplier No. PEP1475
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic Peptide

This peptide corresponds to 18 amino acid peptide near the carboxy terminus of human DLK1. PEP-1475 can be used as a blocking peptide with polyclonal antibody PA5-34433.

The Delta-like 1 homolog (DLK1) is a transmembrane protein containing six epidermal growth factor repeats. It is involved in the differentiation of several cell types, including adipocytes and is also thought to be a tumor suppressor. The DLK1 gene is one of several imprinted genes located in a region on chromosome 14q32; certain mutations in this imprinted region can cause phenotypes similar to maternal and paternal uniparental disomy of chromosome 14 (UPD14). DLK1 is expressed from the paternal allele; a polymorphism within this gene has been associated with child and adolescent obesity.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 8788
Species Human
Purification Method purified
Accession Number P80370
Common Name DLK1
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias Adipocyte differentiation inhibitor protein; AW742678; delta like non-canonical Notch ligand 1; Delta1; delta-like 1 homolog; delta-like 1 homolog (Drosophila); DLK; Dlk1; DLK-1; DlkI; FA1; Fetal antigen 1; Ly107; Peg9; pG2; Preadipocyte factor 1; preadipocyte factor-1; PREF; Pref1; pref-1; Protein delta homolog 1; SCP1; Scp-1; secredeltin; stromal cell derived protein 1; ZOG
Gene Symbol DLK1
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.