Learn More
ACROBiosystems Biotinylated Human DLL1, His,Avitag (DL1-H82E5) is expressed from human 293 cells (HEK293). It contains AA Gln 18 - Gly 540 (Accession # O00548-1).
Catalog No. 502254036
Encompass_Preferred
Delta-like protein 1 (DLL1) is also known as Drosophila Delta homolog 1 (Delta1 or H-Delta-1), which contains one DSL domain and eight EGF-like domains. DLL1 is ubiquitinated by MIB (MIB1 or MIB2), leading to its endocytosis and subsequent degradation. As for expression, DLL1 is expressed in heart and pancreas, with lower expression in brain and muscle and almost no expression in placenta, lung, liver and kidney. Furthermore, DLL1 acts as a ligand for Notch receptors. Also, DLL1 can block the differentiation of progenitor cells into the B-cell lineage while promoting the emergence of a population of cells with the characteristics of a T-cell/NK-cell precursor.
This item is not returnable. View return policy
Catalog No. 50-225-4036 Supplier ACROBiosystems Supplier No. DL1H82E525UG
Delta-like protein 1 (DLL1) is also known as Drosophila Delta homolog 1 (Delta1 or H-Delta-1), which contains one DSL domain and eight EGF-like domains. DLL1 is ubiquitinated by MIB (MIB1 or MIB2), leading to its endocytosis and subsequent degradation. As for expression, DLL1 is expressed in heart and pancreas, with lower expression in brain and muscle and almost no expression in placenta, lung, liver and kidney. Furthermore, DLL1 acts as a ligand for Notch receptors. Also, DLL1 can block the differentiation of progenitor cells into the B-cell lineage while promoting the emergence of a population of cells with the characteristics of a T-cell/NK-cell precursor.