Learn More
Sino Biological Anti-DLL1 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 11635MM0720
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human DLL1 / Delta-1 (rh DLL1; Catalog#11635-H08H; NP_005609.3; Met 1-Gly 540). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-1749