Learn More
Sino Biological Recombinant Human DLL1 Protein (ECD, hFc Tag), HPLC-verified
Supplier:  Sino Biological 11635H02H50
Encompass_Preferred
A DNA sequence encoding the human DLL1 (NP_005609.3) (Met1-Gly540) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-255-4656