Learn More
ACROBiosystems ActiveMax Human DLL3 μBeads, premium grade (for cells)
Catalog No. 502719179
Encompass_Preferred
ActiveMax Human DLL3 μBeads, premium grade (for cells) are produced under sterile manufacturing conditions (ISO 5) , and no animal- or human-derived components are used throughout the production process. It is produced under our rigorous quality control system that includes a comprehensive set of tests including sterility and endotoxin tests.
Catalog No. 50-271-9179 Supplier ACROBiosystems Supplier No. MBSC0182.5MG
ActiveMax Human DLL3 μBeads, premium grade (for cells) are produced under sterile manufacturing conditions (ISO 5) , and no animal- or human-derived components are used throughout the production process. It is produced under our rigorous quality control system that includes a comprehensive set of tests including sterility and endotoxin tests.