Learn More
ACROBiosystems Human DLL3 200tests
Catalog No. 502535176
Encompass_Preferred
APC-Labeled Human DLL3 Protein His Tag (DL3-HA2H8) is produced via conjugation of APC to Human DLL3 Protein His Tag with a new generation site-specific technology under Star Staining labeling platform. Human DLL3 Protein His Tag is is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Leu 492 (Accession # Q9NYJ7-1).
Catalog No. 50-253-5176 Supplier ACROBiosystems Supplier No. DL3HA2H8200TESTS
APC-Labeled Human DLL3 Protein His Tag (DL3-HA2H8) is produced via conjugation of APC to Human DLL3 Protein His Tag with a new generation site-specific technology under Star Staining labeling platform. Human DLL3 Protein His Tag is is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Leu 492 (Accession # Q9NYJ7-1).