Learn More
ACROBiosystems PE-Labeled Human DLL3 Protein, His Tag (Site-specific conjugation)
Supplier:  ACROBiosystems DL3HP2H7200TESTS
Encompass_Preferred
PE-Labeled Human DLL3 Protein, His Tag (DL3-HP2H7) is produced via site-specific conjugation of PE to Human DLL3 Protein, His Tag under optimal conditions with a proprietary technology. Human DLL3 Protein, His Tag is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Leu 492 (Accession # Q9NYJ7-1).
Catalog No. 50-233-1195