Learn More
ACROBiosystems Human DLL3 25ug
Catalog No. 502535177
Encompass_Preferred
Alexa Fluor 647-Labeled Human DLL3 Protein His Tag (DL3-HA2H3) is produced via conjugation of AF647 to Human DLL3 Protein His Tag with a new generation site-specific technology under Star Staining labeling platform. Human DLL3 Protein His Tag is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Leu 492 (Accession # Q9NYJ7-1).
Catalog No. 50-253-5177 Supplier ACROBiosystems Supplier No. DL3HA2H325UG
Alexa Fluor 647-Labeled Human DLL3 Protein His Tag (DL3-HA2H3) is produced via conjugation of AF647 to Human DLL3 Protein His Tag with a new generation site-specific technology under Star Staining labeling platform. Human DLL3 Protein His Tag is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Leu 492 (Accession # Q9NYJ7-1).