Learn More
ACROBiosystems Human DLL3 ELISA Kit - 96 tests; sandwich ELISA for serum, plasma, and cell culture supernatant
Catalog No. NC3902890
Encompass_Preferred
Human DLL3 ELISA Kit is a sandwich-format immunoassay for quantitative measurement of human DLL3 in biological samples. The ready-to-use 96-test kit contains a pre-coated microplate, standards, detection antibody, streptavidin-HRP, buffers, substrate, and stop solution, and is optimized for sensitivity and reproducibility.
  • Provides quantitative measurement of DLL3 in serum, plasma, and cell culture supernatants.
  • Sandwich ELISA format with pre-coated microplate.
  • Includes standards, biotinylated detection antibody, streptavidin-HRP, wash and dilution buffers, substrate, and stop solution.
  • Validated sensitivity of 25 pg/mL and detection range 54.69-3,500 pg/mL.
  • Supplied for 96 tests.
  • Store unopened kit at 2-8 °C; opened components per label; opened shelf life 30 days.
  • Designed for research use with validated performance for recovery, linearity, and precision.
Catalog No. NC3902890 Supplier ACROBiosystems Supplier No. CEAB04996TESTS
Human DLL3 ELISA Kit is a sandwich-format immunoassay for quantitative measurement of human DLL3 in biological samples. The ready-to-use 96-test kit contains a pre-coated microplate, standards, detection antibody, streptavidin-HRP, buffers, substrate, and stop solution, and is optimized for sensitivity and reproducibility.
  • Provides quantitative measurement of DLL3 in serum, plasma, and cell culture supernatants.
  • Sandwich ELISA format with pre-coated microplate.
  • Includes standards, biotinylated detection antibody, streptavidin-HRP, wash and dilution buffers, substrate, and stop solution.
  • Validated sensitivity of 25 pg/mL and detection range 54.69-3,500 pg/mL.
  • Supplied for 96 tests.
  • Store unopened kit at 2-8 °C; opened components per label; opened shelf life 30 days.
  • Designed for research use with validated performance for recovery, linearity, and precision.