Learn More
ACROBiosystems Human DLL3 (176-492) , His Tag (DL3-H52Hy) is expressed from human 293 cells (HEK293). It contains AA Ala 176 - Leu 492 (Accession # Q9NYJ7-1).
Catalog No. 502230967
Encompass_Preferred
Delta-like protein 3 (DLL3) is a transmembrane protein that belongs to the Delta/Serrate/Lag-2 (DSL) family of Notch ligands.May be required to divert neurons along a specific differentiation pathway. Plays a role in the formation of somite boundaries during segmentation of the paraxial mesoderm. DLL3 protein is expressed on the surface of tumor cells but not in normal adult tissues.
This item is not returnable. View return policy
Catalog No. 50-223-0967 Supplier ACROBiosystems Supplier No. DL3H52HY100UG
Delta-like protein 3 (DLL3) is a transmembrane protein that belongs to the Delta/Serrate/Lag-2 (DSL) family of Notch ligands.May be required to divert neurons along a specific differentiation pathway. Plays a role in the formation of somite boundaries during segmentation of the paraxial mesoderm. DLL3 protein is expressed on the surface of tumor cells but not in normal adult tissues.