Learn More
ACROBiosystems Human DLL3 (312-492) , His Tag (DL3-H52Hz) is expressed from human 293 cells (HEK293). It contains AA Ser 312 - Leu 492 (Accession # Q9NYJ7-1).
Supplier:  ACROBiosystems DL3H52HZ100UG
Encompass_Preferred
Delta-like protein 3 (DLL3) is a transmembrane protein that belongs to the Delta/Serrate/Lag-2 (DSL) family of Notch ligands.May be required to divert neurons along a specific differentiation pathway. Plays a role in the formation of somite boundaries during segmentation of the paraxial mesoderm. DLL3 protein is expressed on the surface of tumor cells but not in normal adult tissues.
Catalog No. 50-223-0969