Learn More
ACROBiosystems Biotinylated Human DLL4, His,Avitag (DL4-H82E6) is expressed from human 293 cells (HEK293). It contains AA Ser 27 - Pro 524 (Accession # NP_061947.1).
Catalog No. 502254127
Encompass_Preferred
Delta-like protein 4 (DLL4) is also known as Drosophila Delta homolog 4 (Delta4), which contains one DSL domain and eight EGF-like domains. DLL4 is expressed in vascular endothelium. DLL4 is involved in the Notch signaling pathway as Notch ligand, which can activates NOTCH1 and NOTCH4. DLL4 is involved in angiogenesis and negatively regulates endothelial cell proliferation and migration and angiogenic sprouting. DLL4 can bind to Notch-1 and Notch-4.
This item is not returnable. View return policy
Catalog No. 50-225-4127 Supplier ACROBiosystems Supplier No. DL4H82E625UG
Delta-like protein 4 (DLL4) is also known as Drosophila Delta homolog 4 (Delta4), which contains one DSL domain and eight EGF-like domains. DLL4 is expressed in vascular endothelium. DLL4 is involved in the Notch signaling pathway as Notch ligand, which can activates NOTCH1 and NOTCH4. DLL4 is involved in angiogenesis and negatively regulates endothelial cell proliferation and migration and angiogenic sprouting. DLL4 can bind to Notch-1 and Notch-4.