Learn More
Sino Biological Recombinant Human DLL4 Protein (His Tag), HPLC-verified 1 mg
Supplier:  Sino Biological 10171H08H1
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Pro 524) of human DLL4 (NP_061947.1) pro-protein was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-6812