Learn More
Sino Biological Anti-DLL4 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10171MM0620
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human DLL4 / Delta4 extracellular domain (rh DLL4; Catalog#10171-H08H; NP_061947.1; Met 1-Pro 524). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-1493