Learn More
Sino Biological Human DLL4 MAb 100ug (Sino Biological)
Supplier:  Sino Biological 10171R003H100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human DLL4 / Delta4 extracellular domain (rh DLL4; Catalog#10171-H08H; NP_061947.1; Met 1-Pro 524), and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-237-3917