Learn More
BIOHIPPO Human DMKN Microsample
SDP
Catalog No. 5000314465
Encompass
Human DMKN Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of DMKN in serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 056 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human DMKN Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human DMKN Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human DMKN biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14465 Supplier BIOHIPPO Supplier No. BHE15211015
Human DMKN Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of DMKN in serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 056 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human DMKN Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human DMKN Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human DMKN biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color