Learn More
Raybiotech Inc Human DMP-1 ELISA Kit
Supplier:  Raybiotech Inc ELHDMP11
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 80 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detect rhDMP-1, it shows no cross-reactivity with rmDMP-1, rhCD44, or rhComplement Factor H
Catalog No. 50-194-9194