Learn More
American Research Products Inc Human DNA binding protein inhi
Catalog No. 5000327340
Encompass
Human DNA-binding protein inhibitor ID-1 (ID1) ELISA kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-27340 Supplier American Research Products Inc Supplier No. CSBEL010966HU
Human DNA-binding protein inhibitor ID-1 (ID1) ELISA kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich